Merck
  • Metagenomic insight into the microbial networks and metabolic mechanism in anaerobic digesters for food waste by incorporating activated carbon.
Metagenomic insight into the microbial networks and metabolic mechanism in anaerobic digesters for food waste by incorporating activated carbon.
Scientific reports (2017-09-14)
Jingxin Zhang, Liwei Mao, Le Zhang, Kai-Chee Loh, Yanjun Dai, Yen Wah Tong
ABSTRACT
Powdered activated carbon (AC) is commonly used as an effective additive to enhance anaerobic digestion (AD), but little is known about how the metabolic pathways resulting from adding AC change the microbial association network and enhance food waste treatment. In this work, the use of AC in an anaerobic digestion system for food waste was explored. Using bioinformatics analysis, taxonomic trees and the KEGG pathway analysis, changes in microbial network and biometabolic pathways were tracked. The overall effect of these changes were used to explain and validate improved digestion performance. The results showed that AC accelerated the decomposition of edible oil in food waste, enhancing the conversion of food waste to methane with the optimized dosage of 12 g AC per reactor. Specifically, when AC was added, the proponoate metabolic pathway that converts propanoic acid to acetic acid became more prominent, as measured by 16S rRNA in the microbial community. The other two metabolic pathways, Lipid Metabolism and Methane Metabolism, were also enhanced. Bioinformatics analysis revealed that AC promoted the proliferation of syntrophic microorganisms such as Methanosaeta and Geobacter, forming a highly intensive syntrophic microbial network.