Merck
  • Annexin A1 nuclear translocation induces retinal ganglion cell apoptosis after ischemia-reperfusion injury through the p65/IL-1β pathway.
Annexin A1 nuclear translocation induces retinal ganglion cell apoptosis after ischemia-reperfusion injury through the p65/IL-1β pathway.
Biochimica et biophysica acta (2017-04-09)
Yin Zhao, Xing Li, Jieling Gong, Lu Li, Liwen Chen, Lu Zheng, Zhiqi Chen, Jing Shi, Hong Zhang
ABSTRACT
The degeneration of retinal ganglion cells (RGCs) has been identified as a major problem in glaucoma. Previous studies have indicated an association between annexin A1 (ANXA1) and neuronal cell apoptosis, and RGCs apoptosis in acute ischemia-reperfusion was attributed to an increased production of IL-1β. We found that the expression and nuclear translocation of ANXA1 were upregulated in models of acute ischemia-reperfusion in RGCs in vivo. ANXA1 was found to have a promoting effect on the expression of IL-1β in primary cultured RGCs, which could be inhibited by treatment with ANXA1 shRNA or the p65 inhibitor BAY 11-7082. ANXA1 interacted with p65, and recruited it into the nucleus. Chromatin immunoprecipitation assay revealed that ANXA1 accumulated at the IL-1β gene promoter. The reduction of p65 nuclear translocation using a membrane-permeable ANXA1 peptide containing a Ser5Ala mutation led to a decrease in the expression of IL-1β, and acute ischemia-reperfusion induced RGCs apoptosis in vivo. These results indicate that in RGCs, ANXA1 increases IL-1β expression by recruiting p65 to the nucleus, which induces cell apoptosis. The obtained results may help the development of a novel treatment strategy against RGCs apoptosis in acute ischemia-reperfusion injury.