Merck
  • Cutting Edge: Selective Oral ROCK2 Inhibitor Reduces Clinical Scores in Patients with Psoriasis Vulgaris and Normalizes Skin Pathology via Concurrent Regulation of IL-17 and IL-10.
Cutting Edge: Selective Oral ROCK2 Inhibitor Reduces Clinical Scores in Patients with Psoriasis Vulgaris and Normalizes Skin Pathology via Concurrent Regulation of IL-17 and IL-10.
Journal of immunology (Baltimore, Md. : 1950) (2017-04-09)
Alexandra Zanin-Zhorov, Jonathan M Weiss, Alissa Trzeciak, Wei Chen, Jingya Zhang, Melanie S Nyuydzefe, Carmen Arencibia, Seetharam Polimera, Olivier Schueller, Judilyn Fuentes-Duculan, Kathleen M Bonifacio, Norma Kunjravia, Inna Cueto, Jennifer Soung, Roy M Fleischmann, Alan Kivitz, Mark Lebwohl, Margarita Nunez, Johnnie Woodson, Shondra L Smith, Robert F West, Mark Berger, James G Krueger, John L Ryan, Samuel D Waksal
ABSTRACT
Targeted inhibition of Rho-associated kinase (ROCK)2 downregulates the proinflammatory T cell response while increasing the regulatory arm of the immune response in animals models of autoimmunity and Th17-skewing human cell culture in vitro. In this study, we report that oral administration of a selective ROCK2 inhibitor, KD025, reduces psoriasis area and severity index scores by 50% from baseline in 46% of patients with psoriasis vulgaris, and it decreases epidermal thickness as well as T cell infiltration in the skin. We observed significant reductions of IL-17 and IL-23, but not IL-6 and TNF-α, whereas IL-10 levels were increased in peripheral blood of clinical responders after 12 wk of treatment with KD025. Collectively, these data demonstrate that an orally available selective ROCK2 inhibitor downregulates the Th17-driven autoimmune response and improved clinical symptoms in psoriatic patients via a defined molecular mechanism that involves concurrent modulation of cytokines without deleterious impact on the rest of the immune system.