Merck
  • Dual histone reader ZMYND8 inhibits cancer cell invasion by positively regulating epithelial genes.
Dual histone reader ZMYND8 inhibits cancer cell invasion by positively regulating epithelial genes.
The Biochemical journal (2017-04-23)
Moitri Basu, Isha Sengupta, Md Wasim Khan, Dushyant Kumar Srivastava, Partha Chakrabarti, Siddhartha Roy, Chandrima Das
ABSTRACT
Enhanced migratory potential and invasiveness of cancer cells contribute crucially to cancer progression. These phenotypes are achieved by precise alteration of invasion-associated genes through local epigenetic modifications which are recognized by a class of proteins termed a chromatin reader. ZMYND8 [zinc finger MYND (myeloid, Nervy and DEAF-1)-type containing 8], a key component of the transcription regulatory network, has recently been shown to be a novel reader of H3.1K36Me2/H4K16Ac marks. Through differential gene expression analysis upon silencing this chromatin reader, we identified a subset of genes involved in cell proliferation and invasion/migration regulated by ZMYND8. Detailed analysis uncovered its antiproliferative activity through BrdU incorporation, alteration in the expression of proliferation markers, and cell cycle regulating genes and cell viability assays. In addition, performing wound healing and invasion/migration assays, its anti-invasive nature is evident. Interestingly, epithelial-mesenchymal transition (EMT), a key mechanism of cellular invasion, is regulated by ZMYND8 where we identified its selective enrichment on promoters of