Merck
  • Synthesis, Characterization, and Catalytic Activities of A Nickel(II) Monoamido-Tetradentate Complex: Evidence For Ni
Synthesis, Characterization, and Catalytic Activities of A Nickel(II) Monoamido-Tetradentate Complex: Evidence For Ni
Chemistry (Weinheim an der Bergstrasse, Germany) (2016-12-22)
Kwon Hee Bok, Myoung Mi Lee, Ga Rim You, Hye Mi Ahn, Ka Young Ryu, Sung-Jin Kim, Youngmee Kim, Cheal Kim
ABSTRACT
A new mononuclear nickel(II) complex, [Ni
Sigma-Aldrich
3-Chloroperbenzoic acid, ≤77%
Sigma-Aldrich
2-Cyclohexen-1-ol, 95%
Sigma-Aldrich
8-Aminoquinoline, 98%