Merck
  • Adiponectin association with T-cadherin protects against neointima proliferation and atherosclerosis.
Adiponectin association with T-cadherin protects against neointima proliferation and atherosclerosis.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2017-01-08)
Yuya Fujishima, Norikazu Maeda, Keisuke Matsuda, Shigeki Masuda, Takuya Mori, Shiro Fukuda, Ryohei Sekimoto, Masaya Yamaoka, Yoshinari Obata, Shunbun Kita, Hitoshi Nishizawa, Tohru Funahashi, Barbara Ranscht, Iichiro Shimomura
ABSTRACT
Adiponectin, an adipocyte-derived protein abundant in the circulation, is thought to be protective against atherosclerosis. However, it is not fully understood how the association of adiponectin with vascular cells and its antiatherogenic effect are connected. In this study, T-cadherin was essential for accumulation of adiponectin in the neointima and atherosclerotic plaque lesions, and the adiponectin-T-cadherin association protected against vascular injury. In the apolipoprotein E-knockout (ApoE-KO) mice, adiponectin and T-cadherin colocalized on endothelial cells and synthetic smooth muscle cells in the aortic intima. Notably, aortic adiponectin protein disappeared in T-cadherin/ApoE double-knockout (Tcad/ApoE-DKO) mice with significant elevation of blood adiponectin concentration. Furthermore, in Tcad/ApoE-DKO mice, carotid artery ligation resulted in a significant increase of neointimal thickness compared with ApoE-KO mice. Finally, on a high-cholesterol diet, Tcad/ApoE-DKO mice increased atherosclerotic plaque formation, despite a 5-fold increase in plasma adiponectin level compared with that in ApoE-KO mice.