Merck
  • Efficacy of phosphodiesterase-4 inhibitors in juvenile Batten disease (CLN3).
Efficacy of phosphodiesterase-4 inhibitors in juvenile Batten disease (CLN3).
Annals of neurology (2016-11-03)
Amy Aldrich, Megan E Bosch, Rachel Fallet, Jessica Odvody, Maria Burkovetskaya, Kakulavarapu V Rama Rao, Jonathan D Cooper, Arlene V Drack, Tammy Kielian
ABSTRACT
Juvenile neuronal ceroid lipofuscinosis (JNCL), or juvenile Batten disease, is a pediatric lysosomal storage disease caused by autosomal recessive mutations in CLN3, typified by blindness, seizures, progressive cognitive and motor decline, and premature death. Currently, there is no treatment for JNCL that slows disease progression, which highlights the need to explore novel strategies to extend the survival and quality of life of afflicted children. Cyclic adenosine monophosphate (cAMP) is a second messenger with pleiotropic effects, including regulating neuroinflammation and neuronal survival. Here we investigated whether 3 phosphodiesterase-4 (PDE4) inhibitors (rolipram, roflumilast, and PF-06266047) could mitigate behavioral deficits and cell-specific pathology in the Cln3 In a randomized, blinded study, wild-type (WT) and Cln3 cAMP levels were significantly reduced in the Cln3 These studies reveal neuroprotective effects for PDE4 inhibitors in Cln3
Sigma-Aldrich
Rolipram, solid, ≥98% (HPLC)