Merck
  • Immune Activation Influences SAMHD1 Expression and Vpx-mediated SAMHD1 Degradation during Chronic HIV-1 Infection.
Immune Activation Influences SAMHD1 Expression and Vpx-mediated SAMHD1 Degradation during Chronic HIV-1 Infection.
Scientific reports (2016-12-07)
Weihui Fu, Chao Qiu, Mingzhe Zhou, Lingyan Zhu, Yu Yang, Chenli Qiu, Linxia Zhang, Xuan Xu, Ying Wang, Jianqing Xu, Xiaoyan Zhang
ABSTRACT
SAMHD1 restricts human immunodeficiency virus type 1 (HIV-1) replication in myeloid cells and CD4+ T cells, while Vpx can mediate SAMHD1 degradation to promote HIV-1 replication. Although the restriction mechanisms of SAMHD1 have been well-described, SAMHD1 expression and Vpx-mediated SAMHD1 degradation during chronic HIV-1 infection were poorly understood. Flow cytometric analysis was used to directly visualize ex vivo, and after in vitro SIV-Vpx treatment, SAMHD1 expression in CD4+ T cells and monocytes. Here we report activated CD4+ T cells without SAMHD1 expression were severely reduced, and SAMHD1 in CD4+ T cells became susceptible to SIV-Vpx mediated degradation during chronic HIV-1 infection, which was absent from uninfected donors. These alterations were irreversible, even after long-term fully suppressive antiretroviral treatment. Although SAMHD1 expression in CD4+ T cells and monocytes was not found to correlate with plasma viral load, Vpx-mediated SAMHD1 degradation was associated with indicators of immune activation. In vitro assays further revealed that T-cell activation and an upregulated IFN-I pathway contributed to these altered SAMHD1 properties. These findings provide insight into how immune activation during HIV-1 infection leads to irreparable aberrations in restriction factors and in subsequent viral evasion from host antiviral defenses.