Merck
  • Entamoeba histolytica uses ferritin as an iron source and internalises this protein by means of clathrin-coated vesicles.
Entamoeba histolytica uses ferritin as an iron source and internalises this protein by means of clathrin-coated vesicles.
International journal for parasitology (2008-10-14)
Fernando López-Soto, Arturo González-Robles, Lizbeth Salazar-Villatoro, Nidia León-Sicairos, Carolina Piña-Vázquez, Eduardo Pérez Salazar, Mireya de la Garza
ABSTRACT
Entamoeba histolytica is a parasitic protozoan that produces dysentery and often reaches the liver, leading to abscess formation. Ferritin is an iron-storage protein that is mainly found in liver and spleen in mammals. The liver contains a plentiful source of iron for amoebae multiplying in that organ, making it a prime target for infection since iron is essential for the growth of this parasite. The aim of this study was to determine whether trophozoites are able to take up ferritin and internalise this protein for their growth in axenic culture. Interaction between the amoebae and ferritin was studied by flow cytometry, confocal laser-scanning microscopy and transmission electron microscopy. Amoebae were viable in iron supplied by ferritin. Trophozoites quickly internalised ferritin via clathrin-coated vesicles, a process that was initiated within the first 2 min of incubation. In 30 min, ferritin was found colocalizing with the LAMP-2 protein at vesicles in the cytosol. The uptake of ferritin was time- temperature- and concentration-dependent, specific and saturated at 46 nM of ferritin. Haemoglobin and holo-transferrin did not compete with ferritin for binding to amoebae. Amoebae cleaved ferritin leading to the production of several different sized fragments. Cysteine proteases of 100, 75 and 50 kDa from amoeba extracts were observed in gels copolymerised with ferritin. For a pathogen such as E. histolytica, the capacity to utilise ferritin as an iron source may well explain its high pathogenic potential in the liver.