Merck
  • Copper affects biochemical and physiological responses of Selenastrum gracile (Reinsch).
Copper affects biochemical and physiological responses of Selenastrum gracile (Reinsch).
Ecotoxicology (London, England) (2016-10-23)
Giseli S Rocha, Christopher C Parrish, Ana T Lombardi, Maria da G G Melão
ABSTRACT
Copper is an essential metal for several physiological and metabolic processes, but a narrow range regulate its effect in phytoplankton cells. It can affect the production of biomolecules and be toxic at concentrations slightly above those required, e.g. decreasing photosynthesis and increasing respiration. The aims of this study were to analyse the changes in growth and chlorophyll a synthesis, and in biochemistry (total carbohydrates, proteins, lipids and fatty acids) of the freshwater microalga Selenastrum gracile after exposure to copper. Exponentially growing cells were exposed to 5 concentrations of free copper ions (Cu2+) ranging from 0.7 (control) to 13 × 10-8 M for up to 120 h. Free Cu2+ ion concentrations were calculated through the chemical equilibrium model MINEQL+. We observed that copper was responsible for a decrease in cell density and an increase in total protein and lipid production, but no effect on total carbohydrates was detected. The increase in phospholipids and sterols and a decrease in saturated fatty acids under copper exposure suggest a change in conformation of the cell membrane, by decreasing its fluidity. We suggest this serves the cell as a system to avoid the internalization of metal, thereby acting as a detoxifying mechanism.
Product Description
Supelco
Bacterial Acid Methyl Ester (BAME) Mix, solution (10 mg/mL total concentration in methyl caproate), analytical standard
Supelco
PUFA No.3, From Menhaden Oil, analytical standard