Merck
  • The oncometabolite 2-hydroxyglutarate activates the mTOR signalling pathway.
The oncometabolite 2-hydroxyglutarate activates the mTOR signalling pathway.
Nature communications (2016-09-15)
Mélissa Carbonneau, Laurence M Gagné, Marie-Eve Lalonde, Marie-Anne Germain, Alena Motorina, Marie-Christine Guiot, Blandine Secco, Emma E Vincent, Anthony Tumber, Laura Hulea, Jonathan Bergeman, Udo Oppermann, Russell G Jones, Mathieu Laplante, Ivan Topisirovic, Kevin Petrecca, Marc-Étienne Huot, Frédérick A Mallette
ABSTRACT
The identification of cancer-associated mutations in the tricarboxylic acid (TCA) cycle enzymes isocitrate dehydrogenases 1 and 2 (IDH1/2) highlights the prevailing notion that aberrant metabolic function can contribute to carcinogenesis. IDH1/2 normally catalyse the oxidative decarboxylation of isocitrate into α-ketoglutarate (αKG). In gliomas and acute myeloid leukaemias, IDH1/2 mutations confer gain-of-function leading to production of the oncometabolite R-2-hydroxyglutarate (2HG) from αKG. Here we show that generation of 2HG by mutated IDH1/2 leads to the activation of mTOR by inhibiting KDM4A, an αKG-dependent enzyme of the Jumonji family of lysine demethylases. Furthermore, KDM4A associates with the DEP domain-containing mTOR-interacting protein (DEPTOR), a negative regulator of mTORC1/2. Depletion of KDM4A decreases DEPTOR protein stability. Our results provide an additional molecular mechanism for the oncogenic activity of mutant IDH1/2 by revealing an unprecedented link between TCA cycle defects and positive modulation of mTOR function downstream of the canonical PI3K/AKT/TSC1-2 pathway.
Sigma-Aldrich
Anti-Akt/PKB Antibody, PH Domain, clone SKB1, clone SKB1, Upstate®, from mouse
Sigma-Aldrich
Octyl-(R)-2HG, ≥98% (HPLC)