Merck
  • Expression of the pituitary stem/progenitor marker GFRα2 in human pituitary adenomas and normal pituitary.
Expression of the pituitary stem/progenitor marker GFRα2 in human pituitary adenomas and normal pituitary.
Pituitary (2014-01-10)
Nestoras Mathioudakis, Ram Sundaresh, Alexandra Larsen, William Ruff, Jennifer Schiller, Hugo Guerrero-Cázares, Peter Burger, Roberto Salvatori, Alfredo Quiñones-Hinojosa
ABSTRACT
Recent studies suggest that adult pituitary stem cells may play a role in pituitary tumorigenesis. We sought to explore whether the Glial cell-line derived neurotrophic factor receptor alpha 2 (GFRα2), a recently described pituitary stem/progenitor marker, might be differentially expressed in pituitary adenomas versus normal pituitary. The expression of GFRα2 and other members of the GFR receptor family (GFRα1, α3, α4) were analyzed using RT-PCR, western blot, and immunohistochemistry in 39 pituitary adenomas, 14 normal pituitary glands obtained at autopsy, and cDNA from 3 normal pituitaries obtained commercially. GFRα2 mRNA was ~2.6 fold under-expressed in functioning adenomas (p < 0.01) and ~3.5 fold over-expressed in non-functioning adenomas (NFAs) (p < 0.05) compared to normal pituitary. Among NFAs, GFRα2 was significantly over-expressed (~5-fold) in the gonadotropinoma subtype only (p < 0.05). GFRα2 protein expression appeared to be higher in most NFAs, although there was heterogeneity in protein expression in this group. GFRα2 protein expression appeared consistently lower in functioning adenomas by IHC and western blot. In normal pituitary, GFRα2 was localized in Rathke's remnant, the putative pituitary stem cell niche, and in corticotropes. Our results suggest that the pituitary stem cell marker GFRα2 is under-expressed in functioning adenomas and over-expressed in NFAs, specifically gonadotropinomas. Further studies are required to elucidate whether over-expression of GFRα2 in gonadotropinomas might play a role in pituitary tumorigenesis.
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein Antibody, clone GA5, clone GA5, Chemicon®, from mouse
Sigma-Aldrich
Anti-S-100 Protein Antibody, clone 15E2E2, clone 15E2E2, Chemicon®, from mouse