Merck
  • A New Approach for Detection Improvement of the Creutzfeldt-Jakob Disorder through a Specific Surface Chemistry Applied onto Titration Well.
A New Approach for Detection Improvement of the Creutzfeldt-Jakob Disorder through a Specific Surface Chemistry Applied onto Titration Well.
Biosensors (2012-01-01)
Caroline Mille, Dominique Debarnot, Willy Zorzi, Benaissa El Moualij, Isabelle Quadrio, Armand Perret-Liaudet, Arnaud Coudreuse, Gilbert Legeay, Fabienne Poncin-Epaillard
ABSTRACT
This work illustrates the enhancement of the sensitivity of the ELISA titration for recombinant human and native prion proteins, while reducing other non-specific adsorptions that could increase the background signal and lead to a low sensitivity and false positives. It is achieved thanks to the association of plasma chemistry and coating with different amphiphilic molecules bearing either ionic charges and/or long hydrocarbon chains. The treated support by 3-butenylamine hydrochloride improves the signal detection of recombinant protein, while surface modification with the 3,7-dimethylocta-2,6-dien-1-diamine (geranylamine) enhances the sensitivity of the native protein. Beside the surface chemistry effect, these different results are associated with protein conformation.