Merck
  • A G-quadruplex DNA structure resolvase, RHAU, is essential for spermatogonia differentiation.
A G-quadruplex DNA structure resolvase, RHAU, is essential for spermatogonia differentiation.
Cell death & disease (2015-01-23)
X Gao, W Ma, J Nie, C Zhang, J Zhang, G Yao, J Han, J Xu, B Hu, Y Du, Q Shi, Z Yang, X Huang, Y Zhang
ABSTRACT
G-quadruplex (G4) DNA and G4 DNA resolvase are involved in a variety of biological processes. To understand the biological function of G4 DNA structures and their resolvases in spermatogenesis, we investigated the distribution of G4 structures in mouse testis and identified their alterations during spermatogenesis. Meanwhile, we studied the function of RNA helicase associated with AU-rich element (RHAU), a G4 DNA resolvase, in spermatogenesis with a germ-cell-specific knockout mouse model. The results showed that the ablation of RHAU in germ cells caused the increase of G4 structures and thus resulted in the decrease of spermatogonial differentiation. c-kit, a spermatogonia differentiation-related gene, contains two G4 DNA motifs on its promoter. We found its expression was significantly downregulated in RHAU conditional knockout testis. A further analysis demonstrated that RHAU directly bound to the G4 structures to activate c-kit expression. We concluded that RHAU regulates spermatogonia differentiation by promoting c-kit expression via directly binding to the G4 DNA motifs c-kit promoter.
Sigma-Aldrich
Anti-BrdU Antibody, clone BU-1, culture supernatant, clone BU-1, Upstate®