Merck
  • Synthesis of trifluoro- or difluoromethylated olefins by regio- and stereocontrolled S(N)2' reactions of gem-difluorinated vinyloxiranes.
Synthesis of trifluoro- or difluoromethylated olefins by regio- and stereocontrolled S(N)2' reactions of gem-difluorinated vinyloxiranes.
The Journal of organic chemistry (2006-04-22)
Hisanori Ueki, Takashi Chiba, Tomoya Kitazume
ABSTRACT
This paper presents a highly stereoselective synthesis of trifluoro- or difluoromethylated olefins via an S(N)2' type fluorination or reductions of gem-difluorinated vinyloxiranes. Their fluorination with HF-Py furnished trifluoromethylated allylic alcohols with exclusive E selectivity. On the other hand, their reduction with DIBAL-H afforded difluoromethylated E-allylic alcohols predominantly, whereas the corresponding Z isomers were formed exclusively by treatment with BH3.THF.
Sigma-Aldrich
Borane tetrahydrofuran complex solution, 1.0 M in THF, contains 0.005 M N-Isopropyl-N-methyl-tert-butylamine as stabilizer