Merck
  • BAX inhibitor-1-associated V-ATPase glycosylation enhances collagen degradation in pulmonary fibrosis.
BAX inhibitor-1-associated V-ATPase glycosylation enhances collagen degradation in pulmonary fibrosis.
Cell death & disease (2014-03-15)
M-R Lee, G-H Lee, H-Y Lee, D-S Kim, M J Chung, Y C Lee, H-R Kim, H-J Chae
ABSTRACT
Endoplasmic reticulum (ER) stress is considered one of the pathological mechanisms of idiopathic pulmonary fibrosis (IPF). Therefore, we examined whether an ER stress regulator, Bax inhibitor-1 (BI-1), regulates collagen accumulation, which is both a marker of fibrosis and a pathological mechanism of fibrosis. The presence of BI-1 inhibited the transforming growth factor-β1-induced epithelial-mesenchymal transition of epithelial pulmonary cells and bleomycin-induced pulmonary fibrosis in a mouse model by enhancing collagen degradation, most likely by enhanced activation of the lysosomal V-ATPase through glycosylation. We also found a correlation between post-translational glycosylation of the V-ATPase and its associated chaperone, calnexin, in BI-1-overexpressing cells. BI-1-induced degradation of collagen through lysosomal V-ATPase glycosylation and the involvement of calnexin were confirmed in a bleomycin-induced fibrosis mouse model. These results highlight the regulatory role of BI-1 in IPF and reveal for the first time the role of lysosomal V-ATPase glycosylation in IPF.