Merck
  • Cilostazol suppresses angiotensin II-induced apoptosis in endothelial cells.
Cilostazol suppresses angiotensin II-induced apoptosis in endothelial cells.
Molecular medicine reports (2016-02-11)
Miao-Qian Shi, Fei-Fei Su, Xuan Xu, Xiong-Tao Liu, Hong-Tao Wang, Wei Zhang, Xue Li, Cheng Lian, Qiang-Sun Zheng, Zhi-Chun Feng
ABSTRACT
Patients with essential hypertension undergo endothelial dysfunction, particularly in the conduit arteries. Cilostazol, a type III phosphodiesterase inhibitor, serves a role in the inhibition of platelet aggregation and it is widely used in the treatment of peripheral vascular diseases. Previous studies have suggested that cilostazol suppresses endothelial dysfunction; however, it remains unknown whether cilostazol protects the endothelial function in essential hypertension. The aim of the present study was to investigate whether, and how, cilostazol suppresses angiotensin II (angII)‑induced endothelial dysfunction. Human umbilical vein endothelial cells (HUVECs) and Sprague Dawley rats were exposed to angII and treated with cilostazol. Endothelial cell apoptosis and function, nitric oxide and superoxide production, phosphorylation (p) of Akt, and caspase‑3 protein expression levels were investigated. AngII exposure resulted in the apoptosis of endothelial cells in vitro and in vivo. In vitro, cilostazol significantly suppressed the angII‑induced apoptosis of HUVECs; however, this effect was reduced in the presence of LY294002, a phosphoinositide 3 kinase (PI3K) inhibitor. Furthermore, cilostazol suppressed the angII‑induced p‑Akt downregulation and cleaved caspase‑3 upregulation. These effects were also alleviated by LY294002. In vivo, cilostazol suppressed the angII‑induced endothelial cell apoptosis and dysfunction. Cilostazol was also demonstrated to partially reduced the angII‑induced increase in superoxide production. The results of the present study suggested that cilostazol suppresses endothelial apoptosis and dysfunction by modulating the PI3K/Akt pathway.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-AKT antibody produced in rabbit, affinity isolated antibody