Merck
  • Novel N-substituted 3 alpha-[bis(4'-fluorophenyl)methoxy]tropane analogues: selective ligands for the dopamine transporter.
Novel N-substituted 3 alpha-[bis(4'-fluorophenyl)methoxy]tropane analogues: selective ligands for the dopamine transporter.
Journal of medicinal chemistry (1998-01-22)
G E Agoston, J H Wu, S Izenwasser, C George, J Katz, R H Kline, A H Newman
ABSTRACT
A series of N-substituted 3 alpha-[bis(4'-fluorophenyl)methoxy]tropane analogues has been prepared that function as dopamine uptake inhibitors. The N-methylated analogue of this series had a significantly higher affinity for the dopamine transporter than the parent compound, N-methyl-3 alpha- (diphenylmethoxy)tropane (benztropine, Cogentin). Yet like the parent compound, it retained high affinity for muscarinic receptors. A series of N-substituted compounds were prepared from nor-3 alpha-[bis(4'-fluorophenyl)methoxy]tropane via acylation followed by hydride reduction of the amide or by direct alkylation. All compounds containing a basic tropane nitrogen displaced [3H]WIN 35,428 at the dopamine transporter (Ki range = 8.5-634 nM) and blocked dopamine uptake (IC50 range = 10-371 nM) in rat caudate putamen, whereas ligands with a nonbasic nitrogen were virtually inactive. None of the compounds demonstrated high binding affinity at norepinephrine or serotonin transporters. Importantly, a separation of binding affinities for the dopamine transporter versus muscarinic m1 receptors was achieved by substitution of the N-methyl group with other N-alkyl or arylalkyl substituents (eg. n-butyl, allyl, benzyl, 3-phenylpropyl, etc.). Additionally, the most potent and selective analogue in this series at the dopamine transporter, N-(4"-phenyl-n-butyl)-3 alpha-[bis(4'-fluorophenyl)methoxy]tropane analogue failed to substitute for cocaine in rats trained to discriminate cocaine from saline. Potentially, new leads toward the development of a pharmacotherapeutic for cocaine abuse and other disorders affecting the dopamine transporter may be discovered.