Merck
  • Somatic embryogenesis and massive shoot regeneration from immature embryo explants of tef.
Somatic embryogenesis and massive shoot regeneration from immature embryo explants of tef.
Biotechnology research international (2011-10-27)
Likyelesh Gugsa, Jochen Kumlehn
ABSTRACT
Tef (Eragrostis tef) provides a major source of human nutrition in the Horn of Africa, but biotechnology has had little impact on its improvement to date. Here, we report the elaboration of an in vitro regeneration protocol, based on the use of immature zygotic embryos as explant. Explant size was an important determinant of in vitro regeneration efficiency, as was the formulation of the culture medium. Optimal results were obtained by culturing 0.2-0.35 mm embryo explants on a medium containing KBP minerals, 9.2-13.8 μM 2,4-dichlorophenoxyacetic acid, 6 mM glutamine, and 0.5% Phytagel. Although this protocol was effective for both the improved cultivar "DZ-01-196" and the landrace "Fesho", the former produced consistently more embryogenic tissue and a higher number of regenerants. An average of more than 2,800 shoots could be obtained from each "DZ-01-196" explant after 12 weeks of in vitro culture. These shoots readily formed roots, and plantlets transferred to soil were able to develop into morphologically normal, fertile plants. This regeneration and multiplication system should allow for the application of a range of biotechnological methods to tef.
Sigma-Aldrich
Kao and Michayluk Vitamin Solution, 100 ×, liquid, suitable for plant cell culture
Pricing and availability is not currently available.