Merck
  • Versican is a potential therapeutic target in docetaxel-resistant prostate cancer.
Versican is a potential therapeutic target in docetaxel-resistant prostate cancer.
Oncoscience (2015-04-11)
Naoko Arichi, Yozo Mitsui, Miho Hiraki, Sigenobu Nakamura, Takeo Hiraoka, Masahiro Sumura, Hiroshi Hirata, Yuichiro Tanaka, Rajvir Dahiya, Hiroaki Yasumoto, Hiroaki Shiina
ABSTRACT
In the current study, we investigated a combination of docetaxel and thalidomide (DT therapy) in castration-resistant prostate cancer (CRPC) patients. We identified marker genes that predict the effect of DT therapy. Using an androgen-insensitive PC3 cell line, we established a docetaxel-resistant PC-3 cell line (DR-PC3). In DR-PC3 cells, DT therapy stronger inhibited proliferation/viability than docetaxel alone. Based on gene ontology analysis, we found versican as a selective gene. This result with the findings of cDNA microarray and validated by quantitative RT-PCR. In addition, the effect of DT therapy on cell viability was the same as the effect of docetaxel plus versican siRNA. In other words, silencing of versican can substitute for thalidomide. In the clinical setting, versican expression in prostate biopsy samples (before DT therapy) correlated with PSA reduction after DT therapy (p<0.05). Thus targeting versican is a potential therapeutic strategy in docetaxel-resistant prostate cancer.