Merck
  • Interaction of cytosolic adaptor proteins with neuronal apolipoprotein E receptors and the amyloid precursor protein.
Interaction of cytosolic adaptor proteins with neuronal apolipoprotein E receptors and the amyloid precursor protein.
The Journal of biological chemistry (1998-12-05)
M Trommsdorff, J P Borg, B Margolis, J Herz
ABSTRACT
Apolipoprotein E, alpha2-macroglobulin, and amyloid precursor protein (APP) are involved in the development of Alzheimer's disease. All three proteins are ligands for the low density lipoprotein (LDL) receptor-related protein (LRP), an abundant neuronal surface receptor that has also been genetically linked to Alzheimer's disease. The cytoplasmic tails of LRP and other members of the LDL receptor gene family contain NPxY motifs that are required for receptor endocytosis. To investigate whether these receptors may have functions that go beyond ligand internalization, e.g. possible roles in cellular signaling, we searched for proteins that might interact with the cytoplasmic tails of the receptors. A family of adaptor proteins containing protein interaction domains that can interact with NPxY motifs has previously been described. Using yeast 2-hybrid and protein coprecipitation approaches in vitro, we show that the neuronal adaptor proteins FE65 and mammalian Disabled bind to the cytoplasmic tails of LRP, LDL receptor, and APP, where they can potentially serve as molecular scaffolds for the assembly of cytosolic multiprotein complexes. FE65 contains two distinct protein interaction domains that interact with LRP and APP, respectively, raising the possibility that LRP can modulate the intracellular trafficking of APP. Tyrosine-phosphorylated mammalian Disabled can recruit nonreceptor tyrosine kinases, such as src and abl, to the cytoplasmic tails of the receptors to which it binds, suggesting a molecular pathway by which receptor/ligand interaction on the cell surface could generate an intracellular signal.