Merck
  • Synthesis and antihyperglycemic activity of novel 5-(naphthalenylsulfonyl)-2,4-thiazolidinediones.
Synthesis and antihyperglycemic activity of novel 5-(naphthalenylsulfonyl)-2,4-thiazolidinediones.
Journal of medicinal chemistry (1990-05-01)
A Zask, I Jirkovsky, J W Nowicki, M L McCaleb
ABSTRACT
A series of 5-(naphthalenylsulfonyl)-2,4-thiazolidinediones were synthesized and evaluated for antihyperglycemic activity in an insulin-resistant, genetically diabetic db/db mouse model of non-insulin-dependent diabetes mellitus (NIDDM). The sulfones could be synthesized by a novel, selective C-5 sulfonylation of dilithio-2,4-thiazolidinedione with appropriate sulfonyl chlorides. Within this series, naphthalene was found to be superior to other groups for eliciting antihyperglycemic activity, including the p-alkoxyphenyl group found in ciglitazone, a prototypical agent for this activity. Attachment of the 5-sulfonyl-2,4-thiazolidinedione moiety to the 2-naphthalene position led to optimum activity. Other linkers between the naphthalene and 2,4-thiazolidinedione rings, such as thio, methylene, oxy, and sulfinyl led to decreased antihyperglycemic activity. The best analogue, 5-(2-naphthalenylsulfonyl)-2,4-thiazolidinedione (AY-31,637) was equipotent to ciglitazone in two animal models of NIDDM.
Sigma-Aldrich
2,4-Thiazolidinedione, technical grade, 90%
Pricing and availability is not currently available.