Merck
  • VPS53 mutations cause progressive cerebello-cerebral atrophy type 2 (PCCA2).
VPS53 mutations cause progressive cerebello-cerebral atrophy type 2 (PCCA2).
Journal of medical genetics (2014-03-01)
Miora Feinstein, Hagit Flusser, Tally Lerman-Sagie, Bruria Ben-Zeev, Dorit Lev, Orly Agamy, Idan Cohen, Rotem Kadir, Sara Sivan, Esther Leshinsky-Silver, Barak Markus, Ohad S Birk
ABSTRACT
Progressive cerebello-cerebral atrophy (PCCA) leading to profound mental retardation, progressive microcephaly, spasticity and early onset epilepsy, was diagnosed in four non-consanguineous apparently unrelated families of Jewish Moroccan ancestry. Common founder mutation(s) were assumed. Genome-wide linkage analysis and whole exome sequencing were done, followed by realtime PCR and immunofluorescent microscopy. Genome-wide linkage analysis mapped the disease-associated gene to 0.5 Mb on chromosome 17p13.3. Whole exome sequencing identified only two mutations within this locus, which were common to the affected individuals: compound heterozygous mutations in VPS53, segregating as expected for autosomal recessive heredity within all four families, and common in Moroccan Jews (∼1:37 carrier rate). The Golgi-associated retrograde protein (GARP) complex is involved in the retrograde pathway recycling endocytic vesicles to Golgi; c.2084A>G and c.1556+5G>A VPS53 founder mutations are predicted to affect the C-terminal domain of VPS53, known to be critical to its role as part of this complex. Immunofluorescent microscopy demonstrated swollen and abnormally numerous CD63 positive vesicular bodies, likely intermediate recycling/late endosomes, in fibroblasts of affected individuals. Autosomal recessive PCCA type 2 is caused by VPS53 mutations.