Merck
  • N-cadherin, a novel prognostic biomarker, drives malignant progression of colorectal cancer.
N-cadherin, a novel prognostic biomarker, drives malignant progression of colorectal cancer.
Molecular medicine reports (2015-05-06)
Xuebing Yan, Leilei Yan, Sihong Liu, Zezhi Shan, Yuan Tian, Zhiming Jin
ABSTRACT
Recent studies have indicated that the epithelial-mesenchymal transition (EMT) is a key molecular mechanism involved in the development of colorectal cancer (CRC). N-cadherin is a mesenchymal marker of the EMT and has been closely linked to several human malignancies. However, its role in CRC has remained elusive. In the present study, qRT-PCR and western blot analysis indicated that N-cadherin expression was higher in tumor tissues than in that in their adjacent normal tissues. Immunohistochemical evaluation of N-cadherin and E-cadherin (an epithelial marker of the EMT), indicated that N-cadherin expression was significantly associated with tumor differentiation, tumor size as well as tumor, nodes and metastasis stage. Correlation analysis suggested the expression of N-cadherin was negatively correlated with that of E-cadherin in CRC tissues. Kaplan-Meier analysis indicated that patients with high N-cadherin expression had a significantly lower overall survival and disease-free survival rate than those with low N-cadherin expression, while the opposite was found for E-cadherin. Of note, the present study found that high N-cadherin expression was an independent prognostic factor for CRC. In vitro assays showed that N-cadherin was widely expressed in CRC cell lines and silencing of N-cadherin suppressed the proliferation and migration of the CRC cell line HT-29 by upregulating E-cadherin, suggesting a potential role of N-cadherin in inducing EMT. In conclusion, the present study suggested that N-cadherin has the potential of serving as a novel prognostic predictor and a promising therapeutic target for CRC.
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Boric acid, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Boric acid, 99.999% trace metals basis
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
Hematoxylin, certified by the Biological Stain Commission
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Boric acid, BioReagent, for molecular biology, suitable for cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Boric acid, BioXtra, ≥99.5%
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
o-Xylene, anhydrous, 97%
Sigma-Aldrich
Boric acid, 99.97% trace metals basis
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Boric acid, tablet, 1 g boric acid per tablet
Sigma-Aldrich
Boric acid, suitable for electrophoresis, ≥99.5%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Boric acid-11B, ≥99 atom % 11B
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%