Merck
miRNA-204 drives cardiomyocyte proliferation via targeting Jarid2.
International journal of cardiology (2015-08-25)
Dandan Liang, Jun Li, Yahan Wu, Lixiao Zhen, Changming Li, Man Qi, Lijie Wang, Fangfei Deng, Jian Huang, Fei Lv, Yi Liu, Xiue Ma, Zuoren Yu, Yuzhen Zhang, Yi-Han Chen
ABSTRACT
In mammals, the heart grows by hypertrophy but not proliferation of cardiomyocytes after birth. The paucity of cardiomyocyte proliferation limits cardiac regeneration in a variety of heart diseases. To explore the efficient strategies that drive cardiomyocyte proliferation, we employed in vitro and in vivo models to investigate the function of miRNA-204, which was demonstrated to regulate the proliferation and differentiation of human cardiac progenitor cells in our previous study. miRNA-204 overexpression markedly promoted cardiomyocyte proliferation in both neonatal and adult rat cardiomyocytes in vitro. Transgenic mice with the cardiac-specific overexpression of miRNA-204 exhibited excessive cardiomyocyte proliferation throughout the embryonic and adult stages, leading to a pronounced increase in ventricular mass. Accordingly, the cell cycle regulators, including Cyclin A, Cyclin B, Cyclin D2, Cyclin E, CDC2 and PCNA, were upregulated in miRNA-204 transgenic embryonic hearts. Furthermore, we demonstrated that miRNA-204 directly targeted Jarid2. Knockdown of Jarid2 mimicked the pro-proliferative effect of miRNA-204 overexpression on cultured rat cardiomyocytes, whereas enhanced expression of Jarid2 conferred the myocytes with substantial resistance to proliferation by miRNA-204 overexpression. Our findings identify a conserved role for miRNA-204 in regulating cardiomyocyte proliferation by targeting the Jarid2 signaling pathway.
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Vitamin B12, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
Collagenase from Clostridium histolyticum, suitable for release of physiologically active rat hepatocytes, Type IV, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
Vitamin B12, ≥98%
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
SAFC
BIS-TRIS
SAFC
HEPES
SAFC
HEPES
SAFC
BIS-TRIS
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Cyanocobalamin, meets USP testing specifications
Sigma-Aldrich
Laminin from Engelbreth-Holm-Swarm murine sarcoma basement membrane, 1-2 mg/mL in Tris-buffered saline, 0.2 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Fluoroshield with DAPI, histology mounting medium
Supelco
Cyanocobalamin (B12), analytical standard
Sigma-Aldrich
DAPI, for nucleic acid staining