Merck
  • Effects of calcium chelators on intracellular calcium and excitotoxicity.
Effects of calcium chelators on intracellular calcium and excitotoxicity.
Neuroscience letters (1993-02-19)
J M Dubinsky
ABSTRACT
In an attempt to probe the relationship between excitotoxicity and increases in intracellular calcium ([Ca2+]i), BAPTA-AM and its analogs were applied to cultured hippocampal neurons. Chelation of [Ca2+]i depressed and prolonged transient responses to glutamate and did not effect elevation of [Ca2+]i by prolonged exposure. This explains the inability of the chelators to prevent glutamate-induced toxicity.
Sigma-Aldrich
1,2-Bis(2-amino-5-methylphenoxy)ethane-N,N,N′,N′-tetraacetic acid tetrakis(acetoxymethyl) ester, ≥95% (HPLC)