Merck
  • Simple mass spectrometric method for the estimation of boron and aluminum in water at the parts per billion level.
Simple mass spectrometric method for the estimation of boron and aluminum in water at the parts per billion level.
European journal of mass spectrometry (Chichester, England) (2015-08-27)
Nagasathiya Krishnan, Pachaiappan Raman, Vairamani Mariappanadar
ABSTRACT
The coordinating nature of the hydroxy carboxylic acids, such as tartaric and citric acids, has been utilized for the in-situ formation of anions representing the trivalent elements boron and aluminum and two dianions of the hydroxy acid selected under negative electrospray ionization mass spectral conditions. The abundance of these ions could be used for the quantification of boron and aluminum in water at concentrations ranging from 4.0 ppb to 535.0 ppb. For a period of six months, the validity of this method was tested with citric acid as the coordinating agent. Thus, the developed method offers a simple means for the quantification of boron and aluminum in water by negative electrospray ionization mass spectrometry with a single quadrupole mass spectrometer.
Sigma-Aldrich
Aluminum sulfate, 99.99% trace metals basis
Sigma-Aldrich
Boric acid-11B, ≥99 atom % 11B
Sigma-Aldrich
Boric acid, 99.999% trace metals basis
Sigma-Aldrich
Boric acid, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Boric acid, 99.97% trace metals basis
Sigma-Aldrich
Aluminum sulfate, meets USP testing specifications
Sigma-Aldrich
Boric acid, tablet, 1 g boric acid per tablet
Sigma-Aldrich
Boric acid, suitable for electrophoresis, ≥99.5%
Sigma-Aldrich
Boric acid, BioReagent, for molecular biology, suitable for cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Boric acid, BioXtra, ≥99.5%
Sigma-Aldrich
Boric acid, puriss., meets analytical specification of Ph. Eur., BP, NF, 99.5-100.5%, powder
Sigma-Aldrich
Boric acid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Boric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Boric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., buffer substance, ≥99.8%