Merck
  • Upregulated Polo-Like Kinase 1 Expression Correlates with Inferior Survival Outcomes in Rectal Cancer.
Upregulated Polo-Like Kinase 1 Expression Correlates with Inferior Survival Outcomes in Rectal Cancer.
PloS one (2015-06-06)
T G Tut, S H S Lim, I U Dissanayake, J Descallar, W Chua, W Ng, P de Souza, J-S Shin, C S Lee
ABSTRACT
Human polo-like kinase 1 (PLK1) expression has been associated with inferior outcomes in colorectal cancer. Our aims were to analyse PLK1 in rectal cancer, and its association with clinicopathological variables, overall survival as well as tumour regression to neoadjuvant treatment. PLK1 expression was quantified with immunohistochemistry in the centre and periphery (invasive front) of rectal cancers, as well as in the involved regional lymph nodes from 286 patients. Scores were based on staining intensity and percentage of positive cells, multiplied to give weighted scores from 1-12, dichotomised into low (0-5) or high (6-12). PLK1 scores in the tumour periphery were significantly different to adjacent normal mucosa. Survival analysis revealed that low PLK1 score in the tumour periphery had a hazard ratio of death of 0.59 in multivariate analysis. Other predictors of survival included age, tumour depth, metastatic status, vascular and perineural invasion and adjuvant chemotherapy. There was no statistically significant correlation between PLK1 score and histological tumour regression in the neoadjuvant cohort. Low PLK1 score was an independent predictor of superior overall survival, adjusting for multiple clinicopathological variables including treatment.
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer