Merck
  • Effects of prostacyclin on the early inflammatory response in patients with traumatic brain injury-a randomised clinical study.
Effects of prostacyclin on the early inflammatory response in patients with traumatic brain injury-a randomised clinical study.
SpringerPlus (2014-03-07)
Marie Rodling Wahlström, Magnus Olivecrona, Clas Ahlm, Anders Bengtsson, Lars-Owe D Koskinen, Silvana Naredi, Magnus Hultin
ABSTRACT
A prospective, randomised, double-blinded, clinical trial was performed at a level 1 trauma centre to determine if a prostacyclin analogue, epoprostenol (Flolan®), could attenuate systemic inflammatory response in patients with severe traumatic brain injury (TBI). 46 patients with severe TBI, randomised to epoprostenol (n = 23) or placebo (n = 23). Epoprostenol, 0.5 ng · kg(-1) · min(-1), or placebo (saline) was given intravenously for 72 hours and then tapered off over the next 24 hours. Interleukin-6 (IL-6), interleukin-8 (IL-8), soluble intracellular adhesion molecule-1 (sICAM-1), C-reactive protein (CRP), and asymmetric dimethylarginine (ADMA) levels were measured over five days. Measurements were made at 24 h intervals ≤24 h after TBI to 97-120 h after TBI. A significantly lower CRP level was detected in the epoprostenol group compared to the placebo group within 73-96 h (p = 0.04) and within 97-120 h (p = 0.008) after trauma. IL-6 within 73-96 h after TBI was significantly lower in the epoprostenol group compared to the placebo group (p = 0.04). ADMA was significantly increased within 49-72 h and remained elevated, but there was no effect of epoprostenol on ADMA levels. No significant differences between the epoprostenol and placebo groups were detected for IL-8 or sICAM-1. Administration of the prostacyclin analogue epoprostenol significantly decreased CRP and, to some extent, IL-6 levels in patients with severe TBI compared to placebo. These findings indicate an interesting option for treatment of TBI and warrants future larger studies. ClinicalTrials.gov Identifier, NCT01363583.