Merck
  • Characterization of the Vibrio parahaemolyticus Na+/Glucose cotransporter. A bacterial member of the sodium/glucose transporter (SGLT) family.
Characterization of the Vibrio parahaemolyticus Na+/Glucose cotransporter. A bacterial member of the sodium/glucose transporter (SGLT) family.
The Journal of biological chemistry (2000-06-15)
Z Xie, E Turk, E M Wright
ABSTRACT
The Vibrio parahaemolyticus sodium/glucose transporter (vSGLT) is a bacterial member of the SGLT gene family. Wild-type and mutant vSGLT proteins were expressed in Escherichia coli, and transport activity was measured in intact cells and plasma membrane vesicles. Two cysteine-less vSGLT proteins exhibited sugar transport rates comparable with that of the wild-type protein. Six residues in two regions of vSGLT known to be of functional importance in SGLT1 were replaced individually with cysteine in the cysteine-less protein. Characterization of these single cysteine-substituted vSGLTs showed that two residues (Gly-151 and Gln-428) are essential for transport function, whereas the other four residues (Leu-147, Leu-149, Ala-423, and Gln-425) are not. 2-Aminoethylmethanethiosulfonate (MTSEA) blocked Na(+)/glucose transport by only the transporter bearing a cysteine at position 425 (Q425C). MTSEA inhibition was reversed by dithiothreitol and blocked by the presence of both Na(+) and d-glucose, indicating that conformational changes of the vSGLT protein are involved in Na(+)/glucose transport. A split version of vSGLT was generated by co-expression of the N-terminal (N(7)) and C-terminal (C(7)) halves of the transporter. The split vSGLT maintained Na(+)-dependent glucose transport activity. Chemical cross-linking of split vSGLT, with a cysteine in each N(7) and C(7) fragment, suggested that hydrophilic loops between helices 4 and 5 and between helices 10 and 11 are within 8 A of each other. We conclude that the mechanism of Na(+)/glucose transport by vSGLT is similar to mammalian SGLTs and that further studies on vSGLT will provide novel insight to the structure and function of this class of cotransporters.