Merck
A new method for evaluating the dissolution of orodispersible films.
Pharmaceutical development and technology (2014-02-04)
Yiran Xia, Fang Chen, Huiping Zhang, Chunlin Luo
ABSTRACT
The aim of this research was to develop and assess a new dissolution apparatus for orodispersible films (ODFs). The new apparatus was based on a flow-through cell design which requires only a limited amount of dissolution medium and can automatically collect samples in short-time intervals. Compared with the dissolution method in Chinese Pharmacopeia, our method simulated the flow condition of the oral cavity and resulted in reproducible dissolution data and remarkably discriminating capability. Therefore, we concluded that the proposed dissolution method was particularly suitable for evaluating the dissolution of ODFs and should also be applicable to other fast-dissolving solid dosage forms.
Sigma-Aldrich
Sodium phosphate monobasic-16O4, 99.9 atom % 16O
Sigma-Aldrich
Sodium phosphate monobasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, BioReagent, for molecular biology, anhydrous, ≥98%
Sigma-Aldrich
Sodium phosphate monobasic, purum p.a., anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium phosphate monobasic, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0% (titration)
Sigma-Aldrich
Sodium phosphate monobasic, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium phosphate monobasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic solution, BioUltra, 5 M in H2O
Sigma-Aldrich
Sodium phosphate monobasic, anhydrous, free-flowing, Redi-Dri, ≥99.0%