Merck
  • The influence of vitamin D analogs on calcification modulators, N-terminal pro-B-type natriuretic peptide and inflammatory markers in hemodialysis patients: a randomized crossover study.
The influence of vitamin D analogs on calcification modulators, N-terminal pro-B-type natriuretic peptide and inflammatory markers in hemodialysis patients: a randomized crossover study.
BMC nephrology (2014-08-13)
Ditte Hansen, Knud Rasmussen, Lars M Rasmussen, Helle Bruunsgaard, Lisbet Brandi
ABSTRACT
The risk of cardiovascular disease is tremendously high in dialysis patients. Dialysis patients treated with vitamin D analogs show decreased cardiovascular morbidity and mortality compared with untreated patients. We examined the influence of two common vitamin D analogs, alfacalcidol and paricalcitol, on important cardiovascular biomarkers in hemodialysis patients. Anti-inflammatory effects and the influence on regulators of vascular calcification as well as markers of heart failure were examined. In 57 chronic hemodialysis patients enrolled in a randomized crossover trial comparing paricalcitol and alfacalcidol, we examined the changes in osteoprotegerin, fetuin-A, NT-proBNP, hs-Crp, IL-6 and TNF-α, during 16 weeks of treatment. NT-proBNP and osteoprotegerin increased comparably in the paricalcitol and alfacalcidol-treated groups. Fetuin-A increased significantly in the alfacalcidol-treated group compared with the paricalcitol-treated group (difference 32.84 μmol/l (95% C.I.; range 0.21-67.47)) during the first treatment period. No difference was found between the groups during the second treatment period, and IL-6, TNF-α and hs-Crp were unchanged in both treatment groups. Paricalcitol and alfacalcidol modulate regulators of vascular calcification. Alfacalcidol may increase the level of the calcification inhibitor fetuin-A. We did not find any anti-inflammatory effect or difference in changes of NT-proBNP. ClinicalTrials.gov NCT00469599 May 3 2007.
Sigma-Aldrich
Mouse Ahsg / Alpha-2-HS-glycoprotein ELISA Kit
Sigma-Aldrich
Human AHSG / Alpha-2-HS-glycoprotein ELISA Kit, for serum, plasma and cell culture supernatants