Merck
  • Haplosufficient genomic androgen receptor signaling is adequate to protect female mice from induction of polycystic ovary syndrome features by prenatal hyperandrogenization.
Haplosufficient genomic androgen receptor signaling is adequate to protect female mice from induction of polycystic ovary syndrome features by prenatal hyperandrogenization.
Endocrinology (2015-02-03)
A S L Caldwell, S Eid, C R Kay, M Jimenez, A C McMahon, R Desai, C M Allan, J T Smith, D J Handelsman, Kirsty A Walters
ABSTRACT
Polycystic ovary syndrome (PCOS) is associated with reproductive, endocrine, and metabolic abnormalities. Because hyperandrogenism is the most consistent PCOS feature, we used wild-type (WT) and androgen receptor (AR) knockout (ARKO) mice, together with a mouse model of PCOS, to investigate the contribution of genomic AR-mediated actions in the development of PCOS traits. PCOS features were induced by prenatal exposure to dihydrotestosterone (250 μg) or oil vehicle (control) on days 16-18 of gestation in WT, heterozygote, and homozygote ARKO mice. DHT treatment of WT mice induced ovarian cysts (100% vs 0%), disrupted estrous cycles (42% vs 100% cycling), and led to fewer corpora lutea (5.0±0.4 vs 9.8±1.8). However, diestrus serum LH and FSH, and estradiol-induced-negative feedback as well as hypothalamic expression of kisspeptin, neurokinin B, and dynorphin, were unaffected by DHT treatment in WT mice. DHT-treated WT mice exhibited a more than 48% increase in adipocyte area but without changes in body fat. In contrast, heterozygous and homozygous ARKO mice exposed to DHT maintained comparable ovarian (histo)morphology, estrous cycling, and corpora lutea numbers, without any increase in adipocyte size. These findings provide strong evidence that genomic AR signaling is an important mediator in the development of these PCOS traits with a dose dependency that allows even AR haplosufficiency to prevent induction by prenatal androgenization of PCOS features in adult life.
Product Description
Europium, wire reel, 50mm, diameter 1.0mm, hard, 99.95%