Merck
  • Serine-arginine protein kinase 1 is associated with breast cancer progression and poor patient survival.
Serine-arginine protein kinase 1 is associated with breast cancer progression and poor patient survival.
Medical oncology (Northwood, London, England) (2014-06-26)
Xing-hua Li, Jun-wei Song, Jun-ling Liu, Shu Wu, Le-shi Wang, Li-yun Gong, Xi Lin
ABSTRACT
Breast cancer is the most common cancer and the second leading cause of mortality for women worldwide. It is necessary to identify valuable molecular markers to predict breast cancer progression in patients and treatment effect. Serine-arginine protein kinase 1 (SRPK1), a member of SR kinase family, phosphorylates the SR splicing factors which plays essential roles in normal cell development and multiple human diseases. In the current study, we wanted to explore if there are any relationships between SRPK1 expression in breast cancer and its clinical characteristics. The results showed that SRPK1 is upregulated in breast cancer cell lines and tissues at both mRNA and protein levels, measured by quantitative reverse transcriptase PCR and Western blotting. Immunohistochemical analysis showed a high expression of SRPK1 in 132 paraffin samples of patients with breast cancer; statistical analyses demonstrated that high expression of SRPK1 significantly correlated with clinical staging of patients with breast cancer (P < 0.001), TNM classification (P < 0.05). Low expression of SRPK1 leads to longer survival time, while high expression of SRPK1 leads to shorter survival time of patients. Multivariate analysis revealed that upregulation of SRPK1 might be an independent prognostic marker for the outcomes of patients with breast cancer. In conclusion, upregulation of SRPK1 might play an important role in the progression of breast cancer and might be considered as the potential diagnostic and therapeutic target to this malignancy.
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, Vetec, reagent grade, ≥98%
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis