Merck
CRR9p polymorphism as a protective factor for lung cancer.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine (2014-06-25)
Yang Chen, Zhiguo Yu, Bo Zhang, Zhengqi Chang, Hong Wang, Zhandi Liu
ABSTRACT
A number of studies have investigated the association between CRR9p polymorphism and risk of lung cancer (LC), yet the role in LC pathogenesis remains unclear owing to inconsistencies across studies. We searched PubMed, Embase, and Web of Science for all medical literature published until January 2014. Pooled odds ratios (ORs) and 95 % confidence intervals (CIs) were obtained by means of the fixed effects model. Data from eight studies satisfying the predesigned inclusion criteria were selected for this meta-analysis. We found a statistically significant evidence for a protective effect on the overall LC risk (TT vs. CC: OR = 0.78, 95 % CI = 0.70-0.87, P het = 0.299; TT vs. CT + CC: OR = 0.81, 95 % CI = 0.73-0.90, P het = 0.113; T vs. C: OR = 0.90, 95 % CI = 0.86-0.95, P het = 0.758; TT + CT vs. CC: OR = 0.92, 95 % CI = 0.87-0.98, P het = 0.892). Both Caucasian and Asian populations were suggested to have a reduced risk of developing such cancer. In the analysis of the association between rs401681 and non-small cell lung cancer (NSCLC) risks, all of the contrast models showed similar results except the CT vs. CC genetic model (OR = 0.93, 95 % CI = 0.84-1.02, P het = 0.568). Our meta-analysis provides supportive evidence that CRR9p polymorphism may influence a risk of LC and NSCLC in a protective model.