Merck
  • Methylation of CpG sites in RNF180 DNA promoter prediction poor survival of gastric cancer.
Methylation of CpG sites in RNF180 DNA promoter prediction poor survival of gastric cancer.
Oncotarget (2014-05-17)
Jingyu Deng, Han Liang, Guoguang Ying, Rupeng Zhang, Baogui Wang, Jun Yu, Daiming Fan, Xishan Hao
ABSTRACT
RNF 180, a novel tumor suppressor, has been implicated in the carcinogenesis and progress of gastric cancer. At present study, we found that lower expression of RNA180 was specific in gastric cancer tissues, and the inconsistently methylated levels of RNF180 promoter were identified in the gastric cancer tissues. Importantly, we demonstrated that the methylated CpG site count and four hypermethylated CpG sites (-116, -80, +97, and +102) were significantly associated with the survival of 400 gastric cancer patients, respectively. With multivariate survival analyses, we demonstrated that both the methylation of combined CpG (-116, -80, +97, and +102) sites and N stage were the independent indictor of prognosis of gastric cancer patients. Eventually, the methylation of combined CpG (-116, -80, +97, and +102) sites was identified to have smaller AIC value than N stage by mean of AIC calculation with the Cox proportional hazards model. These findings indicated that the quantitative detection of RNF180 promoter methylation had the intensive feasibility for evaluation the prognosis of gastric cancer patients in clinic.
Sigma-Aldrich
Sodium bisulfite solution, purum, ~40%