Merck
  • Estrogenic activities of chemicals related to food contact plastics and rubbers tested by the yeast two-hybrid assay.
Estrogenic activities of chemicals related to food contact plastics and rubbers tested by the yeast two-hybrid assay.
Food additives and contaminants (2006-03-21)
Yuko Ogawa, Yoko Kawamura, Chiseko Wakui, Motoh Mutsuga, Tetsuji Nishimura, Kenichi Tanamoto
ABSTRACT
Food contact plastics and rubbers possibly contain many kinds of chemicals such as monomers, oligomers, additives, degradation products of polymers and additives, and impurities. Among them, bisphenol A, nonylphenol, benzylbutyl phthalate, styrene oligomers and hydroxylated benzophenones have been reported to possess estrogenic activities. In this study, other chemicals related to food contact plastics and rubbers, and their metabolites induced by the S9-mixture were tested for their estrogenic activities using the yeast two-hybrid assay. Among the 150 chemicals, 10 chemicals such as bis(4-hydroxyphenyl) methane, 4-cyclohexylphenol, 4-phenylphenol, 4,4'-isopropylidenediphenol alkylphosphite, two type of styrenated phenol (including mono type), tris(nonylphenyl) phosphite, 2,2'-dihydroxy-4-methoxybenzophenone, 2-hydroxy-4-methoxybenzophenone and 2,4-diphenyl-4-methyl-1-pentene, their metabolites and the metabolites of 6 other chemicals, such as 2-(phenylmethyl) phenol, styrenated phenol (di and tri type), 1-(N-phenylamino)naphthalene, 4-tert-butylphenylsalicylate, nonylphenol ethoxylates and 2-methyl-6-tert-butylphenol, displayed estrogenic activities. All of them contained a phenol group in their chemical structures or formed one easily by hydrolysis or metabolism. However, most of the chemicals related to food contact plastics and rubbers, and their metabolites did not show any estrogenicity.
Sigma-Aldrich
4-Phenylphenol, purified by sublimation, 99%