- Differential effects of cdk2 and cdk3 on the control of pRb and E2F function during G1 exit.
Differential effects of cdk2 and cdk3 on the control of pRb and E2F function during G1 exit.
The cyclin-dependent kinases cdk2 and cdk3 are required for the G1-S transition in mammalian cells. Here we show that G1 arrest induced by the corresponding dominant-negative mutants of these enzymes, cdk2dn or cdk3dn, is resistant to the action of SV40 T antigen (T). In the presence of cdk2dn, T released active E2F from negative control by pRb and its related family members (pocket proteins) but failed to induce S-phase. Therefore, among other targets, cdk2 also phosphorylates nonpocket protein substrates in promoting S-phase entry, and T does not mimic all cdk2 functions. In the presence of cdk3dn, however, T failed to induce cell cycle progression or stimulate E2F-dependent transcription activity. Dominant-negative cdk3 inhibited E2F-1, E2F-2, and, less significantly, E2F-3, but not E2F-4 transcription activity. The inhibition occurred in a pRb-independent manner and did not affect the DNA-binding capacity of the transcription factor. Cdk3 bound specifically to E2F-1/DP-1 complexes in vivo, most likely through DP-1. Thus, cdk3 function contributes to the activation of E2F-1, E2F-2, and partially E2F-3 and, thereby, participates in the process of S-phase entry.