Merck
  • Characterization of metabolic phenotypes of mice lacking GPR61, an orphan G-protein coupled receptor.
Characterization of metabolic phenotypes of mice lacking GPR61, an orphan G-protein coupled receptor.
Life sciences (2011-10-06)
Hirohide Nambu, Miyuki Fukushima, Hirohiko Hikichi, Takao Inoue, Norihiro Nagano, Yoshio Tahara, Tadahiro Nambu, Junko Ito, Yoshihiro Ogawa, Satoshi Ozaki, Hisashi Ohta
ABSTRACT
GPR61 is an orphan G protein-coupled receptor whose function remains unknown. The purpose of the present study is to elucidate the importance of GPR61 in metabolism by characterization of GPR61-deficient mice. Male GPR61-deficient mice were characterized regarding various metabolic parameters, including food intake, body weight, oxygen consumption, body temperature, locomotor activity, and in a pair feeding study. Hypothalamic gene expression was analyzed using real-time quantitative RT-PCR. GPR61-deficient mice exhibited marked hyperphagia and heavier body weight than wild-type mice. Hyperphagia of GPR61-deficient mice was observed before the differences in body weight became apparent between the genotypes. When body weight difference did become apparent between genotypes, increases in visceral fat pad weight, liver weight, liver triglyceride (TG) content, plasma leptin, and plasma insulin were observed in GPR61-deficient mice, suggesting that GPR61 deficiency caused obesity associated with hyperphagia. Oxygen consumption, body temperature, and locomotor activity were not significantly different between GPR61-deficient and wild-type mice. Pair-fed GPR61-deficient mice had a greater fat mass than wild-type mice despite comparable body weight in both genotypes. The mRNA levels of proopiomelanocortin (POMC) and brain-derived neurotropic factor (BDNF) in the hypothalamus of GPR61-deficient mice were significantly lower than those of wild-type mice. GPR61-deficient mice exhibited obesity associated with hyperphagia. These findings suggest that GPR61 is involved in the regulation of food intake and body weight, and may be of importance when considering GPR61 as a therapeutic target for obesity or eating disorders.