Merck
Total synthesis of epothilones B and D.
Organic letters (2001-07-07)
R E Taylor, Y Chen
ABSTRACT
[reaction: see text] A highly convergent total synthesis of the natural products epothilone B and D is described. The route is highlighted by efficient generation of a C12-C13 trisubstituted olefin which exploits a sequential Nozaki-Hiyama-Kishi coupling and a stereoselective thionyl chloride rearrangement.
Sigma-Aldrich
6-Heptynoic acid, 90%