Merck
The non-classical export routes: FGF1 and IL-1alpha point the way.
Journal of cell science (2003-11-20)
Igor Prudovsky, Anna Mandinova, Raffaella Soldi, Cinzia Bagala, Irene Graziani, Matteo Landriscina, Francesca Tarantini, Maria Duarte, Stephen Bellum, Holly Doherty, Thomas Maciag
ABSTRACT
Non-classical protein release independent of the ER-Golgi pathway has been reported for an increasing number of proteins lacking an N-terminal signal sequence. The export of FGF1 and IL-1alpha, two pro-angiogenic polypeptides, provides two such examples. In both cases, export is based on the Cu2+-dependent formation of multiprotein complexes containing the S100A13 protein and might involve translocation of the protein across the membrane as a 'molten globule'. FGF1 and IL-1alpha are involved in pathological processes such as restenosis and tumor formation. Inhibition of their export by Cu2+ chelators is thus an effective strategy for treatment of several diseases.