Merck
  • Synthesis and reactivity of an isolable cobalt(I) complex containing a β-diketiminate-based acyclic tetradentate ligand.
Synthesis and reactivity of an isolable cobalt(I) complex containing a β-diketiminate-based acyclic tetradentate ligand.
Inorganic chemistry (2012-02-11)
Elodie E Marlier, Bridget A Ulrich, Kristopher McNeill
ABSTRACT
A model for cobalamin was synthesized using a new monoanionic tetradentate nitrogen donor ligand; 2-(4-tolyl)-1,3-bis(2-isopropylpyridyl)propenediimine (Tol-BDI((2-pp)2)H) (1), which utilizes isopropylpyridines as pendant arms on a β-diketiminate (BDI) backbone. During the synthesis of 1, the rearrangement product, Tol-BDI((2-pp)(4-pp))H (2) was observed. Metalation of 1 with zinc iodide and cobalt chloride yielded the corresponding Tol-BDI((2-pp)2)ZnI (3) and Tol-BDI((2-pp)2)CoCl (4) complexes. The redox properties of 4 in comparison to cobalamin were examined through electrochemical studies. Electrochemical and bulk reduction of complex 4 gave a diamagnetic cobalt(I) complex, Tol-BDI((2-pp)2)Co (5). Reactivity of 5 toward C-X bonds was investigated using methyl iodide and 1-iodo-2-(trimethylsilyl)acetylene, yielding Tol-BDI((2-pp)2)Co(CH(3))I and Tol-BDI((2-pp)2)Co(C(2)Si(CH(3))(3))I respectively. Synthesis and characterization details for these complexes, including the crystal structure of 3, are reported.
Sigma-Aldrich
1-Iodo-2-(trimethylsilyl)acetylene, 97%