Merck
  • HIF-1alpha expression regulates the bactericidal capacity of phagocytes.
HIF-1alpha expression regulates the bactericidal capacity of phagocytes.
The Journal of clinical investigation (2005-07-12)
Carole Peyssonnaux, Vivekanand Datta, Thorsten Cramer, Andrew Doedens, Emmanuel A Theodorakis, Richard L Gallo, Nancy Hurtado-Ziola, Victor Nizet, Randall S Johnson
ABSTRACT
Hypoxia is a characteristic feature of the tissue microenvironment during bacterial infection. Here we report on our use of conditional gene targeting to examine the contribution of hypoxia-inducible factor 1, alpha subunit (HIF-1alpha) to myeloid cell innate immune function. HIF-1alpha was induced by bacterial infection, even under normoxia, and regulated the production of key immune effector molecules, including granule proteases, antimicrobial peptides, nitric oxide, and TNF-alpha. Mice lacking HIF-1alpha in their myeloid cell lineage showed decreased bactericidal activity and failed to restrict systemic spread of infection from an initial tissue focus. Conversely, activation of the HIF-1alpha pathway through deletion of von Hippel-Lindau tumor-suppressor protein or pharmacologic inducers supported myeloid cell production of defense factors and improved bactericidal capacity. HIF-1alpha control of myeloid cell activity in infected tissues could represent a novel therapeutic target for enhancing host defense.
Sigma-Aldrich
3-Hydroxy-1,2-dimethyl-4(1H)-pyridone, 98%