Merck
  • Microwave and flow syntheses of Pseudomonas quinolone signal (PQS) and analogues.
Microwave and flow syntheses of Pseudomonas quinolone signal (PQS) and analogues.
Organic & biomolecular chemistry (2010-10-23)
James T Hodgkinson, Warren R J D Galloway, Shreya Saraf, Ian R Baxendale, Steven V Ley, Mark Ladlow, Martin Welch, David R Spring
ABSTRACT
Expedient syntheses of Pseudomonas quinolone signal (PQS) and related structural analogues using microwave and flow methods are reported.
Sigma-Aldrich
2-Chloro-N-methoxy-N-methylacetamide, 98%