Merck
  • Polyionic polymers--heterogeneous media for metal nanoparticles as catalyst in Suzuki-Miyaura and Heck-Mizoroki reactions under flow conditions.
Polyionic polymers--heterogeneous media for metal nanoparticles as catalyst in Suzuki-Miyaura and Heck-Mizoroki reactions under flow conditions.
Beilstein journal of organic chemistry (2009-07-11)
Klaas Mennecke, Andreas Kirschning
ABSTRACT
The preparation of monolithic polyionic supports which serve as efficient heterogeneous supports for palladium(0) nanoparticles is described. These functionalized polymers were incorporated inside a flow reactor and employed in Suzuki-Miyaura and Heck cross couplings under continuous flow conditions.
Sigma-Aldrich
4′-Iodoacetophenone, ≥97%