Merck
  • Expanded access to naloxone among firefighters, police officers, and emergency medical technicians in Massachusetts.
Expanded access to naloxone among firefighters, police officers, and emergency medical technicians in Massachusetts.
American journal of public health (2014-06-13)
Corey S Davis, Sarah Ruiz, Patrick Glynn, Gerald Picariello, Alexander Y Walley
ABSTRACT
Naloxone is a medication that reverses respiratory depression from opioid overdose if given in time. Paramedics routinely administer naloxone to opioid overdose victims in the prehospital setting, and many states are moving to increase access to the medication. Several jurisdictions have expanded naloxone administration authority to nonparamedic first responders, and others are considering that step. We report here on policy change in Massachusetts, where several communities have equipped emergency medical technicians, law enforcement officers, and firefighters with naloxone.
Product Description
Supelco
Naloxone solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Naloxone for peak identification, European Pharmacopoeia (EP) Reference Standard