Merck
  • Aromatase in breast cancer and the role of aminoglutethimide and other aromatase inhibitors.
Aromatase in breast cancer and the role of aminoglutethimide and other aromatase inhibitors.
Critical reviews in oncology/hematology (1986-01-01)
A M Brodie, R J Santen
ABSTRACT
Approximately one third of human breast carcinomas are hormone dependent and regress upon reduction of circulating estrogen levels. Traditional treatment strategies utilized surgical ablative methods to lower estrogen concentrations as treatment of breast cancer. Currently, investigative emphasis is focused upon development of highly specific antiestrogens and inhibitors of estrogen production. The enzyme, aromatase, as the terminal step in estrogen biosynthesis, is a logical target for blockade with potent and specific inhibitors. The earliest available aromatase antagonist, aminoglutethimide, suppresses estrogen production to the same extent as surgical ablation and is an effective treatment for breast cancer. Aminoglutethimide, however, blocks other cytochrome P-450-mediated steroid hydroxylations, requires concomitant glucocorticoid administration, and is associated with initial side effects. Several more specific inhibitors by destroying aromatase irreversibly as well as by competitive inhibition. One of these, 4-hydroxy-androstenedione, has been intensively studied in animals and is undergoing clinical trial. New data regarding these inhibitors further emphasize the key role of aromatase in estrogen production and the practical utility of blocking this enzyme.
Sigma-Aldrich
DL-Aminoglutethimide
Aminoglutethimide, European Pharmacopoeia (EP) Reference Standard