Merck
  • The all-polyethylene tibial component in primary total knee arthroplasty.
The all-polyethylene tibial component in primary total knee arthroplasty.
The Journal of bone and joint surgery. American volume (2010-02-04)
Terence J Gioe, Aditya V Maheshwari
ABSTRACT
Outcomes of total knee arthroplasties performed with modern all-polyethylene tibial components have been found to be comparable with or better than those of arthroplasties done with metal-backed modular components in numerous mid-to-long-term follow-up studies, radiostereometric analyses, and the few prospective randomized trials available. Advantages of an all-polyethylene tibial component over a metal-backed modular component include lower cost, avoidance of locking-mechanism issues and backside wear, and increased polyethylene thickness after identical bone resections. Disadvantages of an all-polyethylene tibial component compared with a metal-backed modular component include a lack of modularity, limiting intraoperative options; no option for liner removal in the setting of acute irrigation and débridement; and no option for late liner exchange. Primary total knee arthroplasty with a modern all-polyethylene design can be done in many patients, with substantial cost savings across the health-care system.
Product Description
Supelco
Polyethylene, analytical standard, for GPC, 2,000
Sigma-Aldrich
Polyethylene, average Mw ~4,000 by GPC, average Mn ~1,700 by GPC
Sigma-Aldrich
Polyethylene, Medium density
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, average Mw 3,000,000-6,000,000
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, surface-modified, powder, 125 μm avg. part. size
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, surface-modified, powder, 34-50 μm particle size
Sigma-Aldrich
Polyethylene, low density, melt index 25 g/10 min (190°C/2.16kg)
Sigma-Aldrich
Polyethylene, Linear low density, melt index 1.0 g/10 min (190°C/2.16kg)
Sigma-Aldrich
Polyethylene, High density, melt index 12 g/10 min (190 °C/2.16kg)
Sigma-Aldrich
Polyethylene, High density, melt index 2.2 g/10 min (190 °C/2.16kg)
Polyethylene (LDPE), ERM®, certified reference material