Merck
Obesity management: update on orlistat.
Vascular health and risk management (2008-01-19)
Belinda S Drew, Andrew F Dixon, John B Dixon
ABSTRACT
Over the past 20 years obesity has become a worldwide concern of frightening proportion. The World Health Organization estimates that there are over 400 million obese and over 1.6 billion overweight adults, a figure which is projected to almost double by 2015. This is not a disease restricted to adults - at least 20 million children under the age of 5 years were overweight in 2005 (WHO 2006). Overweight and obesity lead to serious health consequences including coronary artery disease, stroke, type-2 diabetes, heart failure, dyslipidemia, hypertension, reproductive and gastrointestinal cancers, gallstones, fatty liver disease, osteoarthritis and sleep apnea (Padwal et al 2003). Modest weight loss in the obese of between 5% and 10% of bodyweight is associated with improvements in cardiovascular risk profiles and reduced incidence of type 2 diabetes (Goldstein 1992; Avenell et al 2004; Padwal and Majumdar 2007). Orlistat, a gastric and pancreatic lipase inhibitor that reduces dietary fat absorption by approximately 30%, has been approved for use for around ten years (Zhi et al 1994; Hauptman 2000). There is now a growing body of evidence to suggest that Orlistat assists weight loss and that it may also have additional benefits. The aim of this review is to provide a brief update on the current literature studying the efficacy, safety and significance of the use of Orlistat in clinical practice.
Product Description
USP
Orlistat, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Orlistat, ≥98%, solid