Merck
  • Alprazolam is relatively more toxic than other benzodiazepines in overdose.
Alprazolam is relatively more toxic than other benzodiazepines in overdose.
British journal of clinical pharmacology (2004-06-23)
Geoffrey K Isbister, Luke O'Regan, David Sibbritt, Ian M Whyte
ABSTRACT
To describe alprazolam poisoning and the relative toxicity of alprazolam compared with other benzodiazepines. A database of consecutive poisoning admissions to a regional toxicology service was searched to identify consecutive benzodiazepine deliberate self poisonings, which were coded as alprazolam, diazepam or other benzodiazepine. Major outcomes used were length of stay (LOS), intensive care (ICU) admission, coma (GCS < 9), flumazenil administration and requirement for mechanical ventilation. Prescription data were obtained for benzodiazepines for the study period. There were 2063 single benzodiazepine overdose admissions: 131 alprazolam overdoses, 823 diazepam overdoses and 1109 other benzodiazepine overdoses. The median LOS for alprazolam overdoses was 19 h which was 1.27 (95% CI 1.04, 1.54) times longer compared with other benzodiazepines by multiple linear regression. For patients with alprazolam overdoses, 22% were admitted to ICU which was 2.06 (95% CI 1.27, 3.33) times more likely compared with other benzodiazepines after multivariate analysis adjusting for age, dose, gender, time to ingestion and co-ingested drugs. Flumazenil was administered to 14% of alprazolam patients and 16% were ventilated, which was significantly more than for other benzodiazepine overdoses (8% and 11%, respectively). Twelve percent of alprazolam overdoses had a GCS < 9 compared with 10% for other benzodiazepines. From benzodiazepine prescription data, total alprazolam prescriptions in Australia increased from 0.13 million in 1992 to 0.41 million in 2001. Eighty five percent of prescriptions were for panic disorder, anxiety, depression or mixed anxiety/depression. Alprazolam was significantly more toxic than other benzodiazepines. The increased prescription of alprazolam to groups with an increased risk of deliberate self poisoning is concerning and needs review.
Product Description
Alprazolam, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Alprazolam
Supelco
Alprazolam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®